VWR®, Beta Radiation Shields
Supplier: VWR
VWR®
113-8820EA 218 EUR
113-8820
VWR®, Beta Radiation Shields
Shields Radiation Shields
Acrylic shield, transparent, thickness: 10 mm, for protection from low energy beta radiation. The dual angled version is angled at both the bottom and top of the shield.
  • Absorb 99,5% of ³²P beta radiation
  • Stable and easy to clean
  • Also protects from splashes
For use on work tables or in fume cupboards.