Culture vessels, cylindrical, with flat flange, Quickfit®
Supplier: SciLabware
FV1L FV2L FV5L FV10L FV20L
201-8553EA 119 EUR
201-8553 201-8555 201-8556 BIBBFV10L BIBBFV20L
Culture vessels, cylindrical, with flat flange, Quickfit®
Reaction Vessels Reaction Vessels, Cylindrical Shape
PYREX®, borosilicate glass.
  • For use with Quickfit® flat flange lids and retaining clips
  • Excellent chemical resistance
  • High temperature resistance
Precision ground, flat flange joint. Ideal for fermentation and culture work.