Funnel holders
Supplier: KARTELL
269
221-1262EA 53.1 EUR
221-1262
Funnel holders
Retort Stands
PP, white, unbreakable.
  • For funnels 50 to 120 mm Ø
  • With bosshead to fit rods 8 to 14 mm Ø
  • Interchangeable insert for different sizes