VWR® Funnel Holder
Supplier: VWR Collection
241-0105EA 143 EUR
241-0105 241-0106
VWR® Funnel Holder
Retort Stands
PP, white.
  • Stainless steel bosshead to fit rods up to 12 mm diameter
  • Acid resistant