Small stand with H-type base (C)
Supplier: VWR Collection
241-0115EA 58.4 EUR
VWRF08545.084
Small stand with H-type base (C)
Stand with H-formed base and steel rod (8×250 mm). The base (125×50 mm) is made of nickel-plated and laquered nodular cast iron.
  • Central hole for rod
  • Robust and stable
  • Good chemical resistance