Square sampling templates
Supplier: Copan
T2905C T2905CS01 T2906C T2906CS01
710-0480EA 63 EUR
710-0480 710-0481 710-0482 710-0483
Square sampling templates
Samplers Sample Dippers
Square sampling templates to define sampling area. For flat surfaces, templates help define the sample size and allows the investigator to determine the number of organisms per cm². This can be used to validate microbial bioburden in cleanrooms and critical areas in which sterile drugs are exposed to the environment and in food processing plants to establish hygiene scores based on colony forming units per cm².
  • For a defined and standardised sampling
  • Two sizes
Informace pro objednávání: Two sizes of sampling templates are available: 4×5 cm (20 cm²) and 10×10 cm (100 cm²).
Balení: Sampling templates in sterile packs, 50 templates/box.