VWR®, Weighing Boats
Supplier: VWR Collection
611-9194EA 55.2 EUR
611-9194 611-9195 611-9196
VWR®, Weighing Boats
Boats Weighing Boats
Borosilicate 3.3 glass.
  • Ideal for weighing small samples
  • Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel