VWR® Parafilm® M, Dispenser for Sealing Film
Supplier: VWR
Parafilm® M
631-0726EA 135 EUR
631-0726
VWR® Parafilm® M, Dispenser for Sealing Film
Sealing Film Dispensers
Acrylic, for clean storage and easy dispensing of Parafilm® M sealing film.
  • For two rolls 50 mm wide or one roll 100 mm wide
  • Serrated blade for easy cutting
  • Easy to clean