Dispenser for Parafilm® M sealing film
Supplier: Brand
Parafilm® M
291-1219EA 187.4 EUR
291-1219
Dispenser for Parafilm® M sealing film
Sealing Film Dispensers
In plastic, light grey. For neat and safe cutting of Parafilm® M sealing film as well as clean storage.
  • For rolls 50 or 100 mm in width
  • Film feed device
  • Safety cutter