Cover spectacles, Armamax AX
Supplier: HONEYWELL SAFETY
1002224 1002221
111-3548EA 9.4 EUR
111-3548 DALL1002221
Cover spectacles, Armamax AX
Safety Glasses
Practical, durable cover glasses (OTG) offering good protection and comfort.
  • Fit comfortably over prescription eyewear
  • Frosted side panels and brow guard reduce glare and reflection
  • Curved sides and raised nose bridge offer a better fit
Weight: 46 g
Weight: 46 g
Certifikace: EN 166; 1F