Safety spectacles, Bandido
Supplier: Bollé Safety
Bandido
111-0977EA 13.7 EUR
111-0977 111-0319
Safety spectacles, Bandido
Safety Glasses
Modern design safety glasses offering a good field of vision.
  • Wrap-around fit
  • Integrated side shields
  • ESP lens coating reduces glare and the harmful effects of blue light (only ESP model)
Weight: 28 g
Certifikace: EN 166, (EN 169, EN 170, EN 172)