Test tube stands
Supplier: KARTELL
1175 1177 1179
212-6201EA 18.8 EUR
212-6201 212-6202 212-6203
Test tube stands
Racks Tube Racks
PS, expanded, white.
  • Good chemical resistance
  • Heat resistant up to 71 °C
  • Extremely light, stackable, with finger recesses for easy lifting
Ideal for large quantities of test and sample tubes. The recesses are deep enough to hold tubes, firm and upright. Cat.No. 212-6201 is particularly suitable for holding 1,5 ml microcentrifuge tubes.