VWR®, Mini Laboratory Notebooks
Supplier: VWR Collection
818-0100EA 34 EUR
818-0100 818-0101 818-0110 818-0111
VWR®, Mini Laboratory Notebooks
Notebooks
Imitation leather for use in laboratory environments. Acid-free, archive safe paper (90 gsm). Includes laboratory notebook guidelines page, table of contents and user data page. English language only.
  • Scientific 5 mm grid or ruled format with date and witness signature blocks
  • ‘FLEX' cover with coating, for durability
Format: A5 (14,8×21 cm) or A6 (10,5×14,8 cm), 96 pages
Colour: blue
All books made in accordance with GLP, GMP and GCP guidelines.