Cleanroom paper, TexWrite® 22
Supplier: Texwipe
TexWrite®
818-0132EA 444 EUR
818-0132
Cleanroom paper, TexWrite® 22
Paper
TexWrite® 22, latex-saturated cellulose cleanroom documentation sheets without organic fillers.
  • Ideal for note taking, equipment manuals, and work instructions
  • Can be used with laser printers and photocopiers
Format: A4 cleanroom bond.
Balení: 250 sheets per pack, 10 packs per case (Bag-Within-A-Bag®).