Wet surface pen, SP Bel-Art
Supplier: Bel-Art
817-1135EA 81.6 EUR
817-1135
Wet surface pen, SP Bel-Art
Pens Ink Pens
It can be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at 0 °C. Can be also used on dry paper as a standard ball-point pen.
  • Writes clearly on wet surfaces