Spore discs, Sportrol™
Supplier: CROSSTEX
BG-106D BS-106D
489-0094EA 199 EUR
489-0094 489-0095
Spore discs, Sportrol™
Sterilisation Indicators
Biological sterilisation discs are used to perform sterilisation cycle validation in small areas of a device, where a standard-sized spore strip will not fit, such as within a syringe barrel, inside tubing, or under a cap. Spore discs are created by inoculating spore suspensions of certified population and resistance onto paper discs.
  • Spore population: 106 spores per disc
  • Disc diameter: 6,35 mm
  • 24-month shelf life when stored under labelled conditions
Certifikace: Meet USP and ISO 11138-1 standards.
Delivery information: Packaged in bulk, in shelfMore Product Information