VWR®, Magnetic stirring bars, cylindrical, with centre ring
Supplier: VWR Collection
442-4500EA 23.8 EUR
442-4500 442-4501 442-4502 442-4503 442-4504 442-4505 442-4506 442-4507 442-4508 442-4509 442-4510 442-4511 442-4512 442-4513 442-0256 442-0257 442-0258
VWR®, Magnetic stirring bars, cylindrical, with centre ring
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. For containers with slightly uneven bases.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)