VWR®, Magnetic Stirring Bars, Plain, Economy
Supplier: VWR Collection
442-0482EA 16.9 EUR
442-0482 442-0483 442-0484 442-0485
VWR®, Magnetic Stirring Bars, Plain, Economy
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges which enhance agitation effect even at low speeds. Inexpensive, high performance.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)