VWR®, Magnetic stirring bars, cylindrical, with removable pivot ring
Supplier: VWR Collection
442-0375EA 45.7 EUR
442-0375 442-0369 442-0371 442-0373 442-0374 442-0376 442-0378 442-0379 442-0381 442-0382 442-0383 442-0384
VWR®, Magnetic stirring bars, cylindrical, with removable pivot ring
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. For containers with slightly uneven bases.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)