VWR®, Magnetic Stirring Bars, Cross-Shaped
Supplier: VWR Collection
442-0394EA 53.2 EUR
442-0394 442-0395 442-0396 442-0397 442-0398 442-0399 442-0400
VWR®, Magnetic Stirring Bars, Cross-Shaped
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Stable rotating position for optimal stirring effect.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)