Magnetic stirring bars, floating, Nalgene®
Supplier: Thermo Fisher Scientific
DS6630-0250 DS6630-1000 DS6630-4000
442-0218EA 111 EUR
442-0218 442-0220 442-0222
Magnetic stirring bars, floating, Nalgene®
Stirrers Magnetic Stirring Bars
Floating stirring bar with PTFE coated Alnico V magnet, which is slightly raised off the bottom of the vessel to minimise grinding effect on cells. Legs can be compressed to fit through small openings.
  • Ideal for low speed stirring action in tissue culture applications
  • Constant action at low speeds
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement