Stoppers for conical ground joints
Supplier: Edwin Mraz Kunststoff-Erzeugnisse GmbH
100/7 100/10 100/12 100/14 10119 100/24 100/29 100/45 100/60
217-8911EA 1.5 EUR
217-8911 217-8912 217-8913 217-8914 217-8915 217-8916 217-8917 217-8918 217-8919
Stoppers for conical ground joints
Stoppers PE/LDPE/HDPE Stoppers
HDPE stopper, white with red cover plate. Hollow, with sealing rings and octagonal head.
  • Good chemical resistance
  • Sealing rings for tight, leak-proof fit
  • Octagonal head allows easy release and removal of the stopper
Certifikace: DIN 12254