Cleanroom tapes
Supplier: ULTRATAPE
1164WH200-P6U 1164WH300-P6U 1164WH100-P6U
817-0072EA 540 EUR
817-0072 817-0073 112-1186
Cleanroom tapes
Tapes
These tapes are made of polyolefin with acrylic adhesive.
  • Ultra low outgassing
  • Low sulfur, low halogen and low heavy metal
  • Adhere lightly to plastics for easy removal
  • Leaves no residue and particulates
Designed specifically for use inside semiconductor and critical environment cleanrooms. It contains extremely low leachable metals, eliminating the concern of contamination.
Length: 54,8 m
Colour: White