Precision thermometers
Supplier: Amarell
L33008-TOLDKD5 L33038-TOL/VC L33058-TOL/CC3 L33008-TOLDKD3 L33004-TOLDKD3 L33000-FL L33000-FL/VC L33000-FL-PTFE L33004-TOL L33008-TOL L33008-TOL/VC L33018-PENT L33018-PENT/VC L33032-FL L33032-FL-PTFE L33032-FL/VC L33038-TOL L33056-TOL L33058-TOL L33070-CB L33074-CB L33076-TOL L33078-TOL L33004-TOL/VC L33004-TOL/DKD L33052-CB/DKD4
AMARL33008-TOLDKD5EA 272.46 EUR
AMARL33008-TOLDKD5 AMARL33038-TOL/VC AMARL33058-TOL/CC3 AMARL33008-TOLDKD3 AMARL33004-TOLDKD3 AMARL33000-FL AMARL33000-FL/VC AMARL33000-FL-PTFE AMARL33004-TOL AMARL33008-TOL AMARL33008-TOL/VC AMARL33018-PENT AMARL33018-PENT/VC AMARL33032-FL AMARL33032-FL-PTFE AMARL33032-FL/VC AMARL33038-TOL AMARL33056-TOL AMARL33058-TOL AMARL33070-CB AMARL33074-CB AMARL33076-TOL AMARL33078-TOL AMARL33004-TOL/VC AMARL33004-TOL/DKD AMARL33052-CB/DKD4
Precision thermometers
Thermometers Analogue Thermometers
This extensive range of low temperature thermometers is suitable for government verification as well as government calibration (620-1713). Toluene or pentane.
  • Total immersion
  • Colourless capillary - except 620-1047 (blue-reflecting capillary)
  • Three different fillings
Ø: 8 - 10 mm
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifikace: Model L33008DKD3 with DIN 12778.
Delivery information: Models 620-1047 and 620-0180 are government verified and supplied with a verification certificate.