VWR®, Thermometer Armor Cases
Supplier: VWR Collection
620-0899EA 23.8 EUR
620-0899 620-0900
VWR®, Thermometer Armor Cases
Thermometer Holders
This case protects your thermometers from accidental breakage during use and storage.
  • Screw cap
  • Ring for hanging
  • Metal or nickel-plated brass
  • Lower section surrounding the bulb with small openings for circulation