Hightech water pump pliers, KNIPEX Cobra®
Supplier: Knipex
238-5550EA 187 EUR
238-5550
Hightech water pump pliers, KNIPEX Cobra®
Pliers
Chrome vanadium electric steel, forged, oil-hardened. For heaviest duty. No more time-consuming presetting of the required opening size. Just position the upper jaw to the workpiece, push button and move close the lower jaw. Chrome plated pliers, handles with non-slip plastic coating.
  • Self-locking on pipes and nuts: no slipping on the workpiece and low handforce required
  • Gripping box-joint design: high stability because of the double guide
  • Reliable catching of the hinge bolt: no unintentional shifting
  • Guard prevents operators' fingers being pinched
Length: 250 mm
Certifikace: DIN ISO 8976