Biodyne® Nylon Transfer Membranes
Supplier: Cytiva (Formerly Pall Lab)
60106 60200 60201 60207 60208 60314 60315 60404 60400 60406 60336 330790
732-3006EA 674 EUR
732-3006 732-3010 732-3011 732-3016 732-3017 732-3018 732-3019 732-3028 732-3024 732-3030 732-2974 514-1437
Biodyne® Nylon Transfer Membranes
Transfer Membranes
Biodyne® Transfer Membranes offer high sensitivity and low background for enhanced detection and resolution. They are ideal for nucleic acid detection.
  • Membranes do not crack, shrink or tear when subjected to multiple cycles of hybridisation, stripping and reprobing
  • Intrinsically hydrophilic for easy wetting
  • Superior performance with radioactive (Biodyne® B membrane) and non-radioactive (Biodyne® A membrane) detection systems
Biodyne® A Membrane (Amphoteric Nylon 6.6): Membrane zeta potential can be modulated by changes in pH. Ideal for single probe or multiple rehybridisations, and applications where background is troublesome.
Biodyne® B Membrane (Positively-charged Nylon 6.6): Pore surfaces are populated by a high density of quaternary ammonium groups. Highest sensitivity for nucleic acid applications.
Biodyne® C Membrane (Negatively-charged Nylon 6.6): Can be derivatised by coupling reactions through the carboxyl groups on the pore surfaces.
Biodyne® Plus Membrane (Positively-charged Nylon 6.6 with high isoelectric point): With certain non-radioactive detection systems, it is more sensitive than Biodyne® A membrane while exhibiting lower background than Biodyne® B membrane. Especially suitable for reverse dot blots and ELISA applications.
Biodyne® Transfer Membranes are resistant to common solvents such as acetone, alcohol, chlorinated aliphatic hydrocarbons, formamide, 2 M sodium hydroxide, DMSO, and dimethylformamide. They are not compatible with concentrated formic acid (>50%), hydrochloric acid (>4 M), oxidising agents, and long exposures (days to weeks) to pH <2.