Centrifuge tubes
Supplier: KARTELL
2300 2302 300 301
212-0209EA 102 EUR
212-0209 212-0284 525-0475 525-0476
Centrifuge tubes
Tubes Centrifuge Tubes
PP, conical bottom, autoclavable.
  • Translucent, excellent chemical resistance
  • Can withstand temperatures up to +120 °C