Centrifuge tubes, Oak Ridge, Nalgene®
Supplier: Thermo Fisher Scientific
3114-0010 3114-0030 3114-0050 3115-0030 3115-0050 3118-0010 3118-0028 3118-0030 3118-0050 3118-0085 3119-0010 3119-0028 3119-0030 3119-0050 3137-0050 3138-0010 3138-0016 3138-0030 3138-0050 3139-0010 3139-0016 3139-0030 3139-0050 3148-0050
525-2211EA 91.2 EUR
525-2211 525-2212 525-2213 525-2223 525-2224 525-2231 525-0037 525-2233 525-2234 525-2235 525-2241 525-0038 525-2243 525-2244 525-0040 525-0041 525-0042 525-0043 525-2236 525-0044 525-0244 525-0045 525-0046 525-0049
Centrifuge tubes, Oak Ridge, Nalgene®
Tubes Centrifuge Tubes
Round bottom and rimless, autoclavable.
  • Withstands forces up to 50000×g
  • Suitable for use in refrigerated or non refrigerated centrifuges
  • At speeds greater than 10000×g, sealing cap assemblies are recommended
FEP
Translucent. Resists any acid, alkali or solvent. Excellent for chloroform and phenol extractions.
PSF
Transparent. Acid and alkali resistant.
PC
Transparent.
PPCO
Translucent. Greater chemical resistance than PC.
Upozornení: Note: (Actual brim capacity of 50 ml Centrifuge tubes Oak Ridge is 42 ml).