Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
T334-2 T334-2S T334-2SPR T334-4 T334-4S T334-4SPR T334-5 T334-5S T334-6 T334-6S T334-7 T334-7S T334-5SPR T334-6SPR T334-7SPR
720-0514EA 240 EUR
720-0514 720-0515 212-9577 212-9578 720-0516 212-9579 212-9580 212-9581 720-0517 720-0518 720-0520 212-9583 212-9582 720-0519 720-0521
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Tubes Microcentrifuge Tubes
PP, with O-ring seal screw cap (without attachment loop), autoclavable.
  • The O-ring secured in the top of the cap ensures a positive leakproof seal, time after time, keeping the integrity of small samples under even the most adverse conditions
  • Sterile tubes are gamma irradiated and available either plain or with printed graduations and white marking area for sample identification
  • Can be autoclaved to +121 °C in upright position with caps loosened
  • Certified RNase-, DNase-, pyrogen- and DNA-free
Ideal for cryogenic work as well as freezer storage, boiling applications, centrifugation etc. and will fit most standard microcentrifuge rotors. The cap's high profile facilitates manipulation especially in aseptic procedures. The cap does not have attachment loops for users who prefer to remove the caps completely from the tubes when filling or sampling.
Max. RCF: Non skirted tubes can be centrifuged up to 20000×g.
Temperature range: –196 to +121 °C.
Balení: Sterile tubes are packaged in tamper-proof resealable bags.