Microcentrifuge tubes, 1,5 ml
Supplier: KARTELL
297 298 298-04 298-06 298-13 298-12
211-2511EA 37.8 EUR
211-2511 211-2512 211-2900 211-2901 211-2902 211-2903
Microcentrifuge tubes, 1,5 ml
Tubes Microcentrifuge Tubes
PP.
  • Ideal for micro-analysis
  • Withstand centrifugation up to 14000×g