PCR microtubes with domed cap, Axygen®
Supplier: Corning
Axygen®
732-1034EA 68.68 EUR
732-1034 525-0248 732-0676
PCR microtubes with domed cap, Axygen®
Tubes PCR Tubes
PP.
  • Domed caps for efficient heat transfer from thermal cycler lid
  • Ultra-thin walls of uniform thickness optimise heat transfer
  • Cap design ensures a tight fit and eliminates possibility of sample evaporation
  • Certified free from DNase, RNase and endotoxin