Test tubes, Corning® Gosselin™
Supplier: Corning
TL75-04 TL75-08 CC21-02
212-5159EA 564 EUR
212-5159 212-5157 GOSSCC21-02
Test tubes, Corning® Gosselin™
Tubes Test Tubes
PS, clear and PP translucent. A range of tubes with conical or round bottoms manufactured under aseptic conditions or sterilised by irradiation.
  • PS tubes crystal clear
  • PP shock resistant and translucent
Suitable for sampling for laboratory testing.
Informace pro objednávání: Available with or without caps.