Test tubes, Corning® Gosselin™
Supplier: Corning
TK75-001 TK75-022 TK75-023 TL40-01 TL40-02 TL40-04 TR95-01 TR95-04 TR95-24 TK75-030 TR95-17 TR95-19 TK75-003 TK75-085 TK75-057
GOSSTK75-001EA 112 EUR
GOSSTK75-001 GOSSTK75-022 GOSSTK75-023 212-5146 212-0164 212-5147 GOSSTR95-01 GOSSTR95-04 GOSSTR95-24 GOSSTK75-030 GOSSTR95-17 GOSSTR95-19 GOSSTK75-003 GOSSTK75-085 GOSSTK75-057
Test tubes, Corning® Gosselin™
Tubes Test Tubes
In transparent PS or PP.
  • Round bottom
  • Aseptic or sterile packaging
  • Sterile tubes are supplied with a cap and non-sterile tubes are supplied without a cap
Universal low volume tubes for a wide range of uses.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904.