VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection
548-0710EA 61.6 EUR
548-0710 548-0711 548-0712 548-0713 548-0723 548-0724 548-0725 548-0726 548-0727 548-0728 548-0729 548-0730 548-0731 548-0732 548-0733 548-0734 548-0735 548-0736 548-0737 548-0714 548-0715 548-0716 548-0717 548-0718 548-0719 548-0720 548-0721 548-0722
VWR® Safe-T-Flex, Caps for Test Tubes
Closures Test/Sample Tube Closures
Safe-T-Flex caps feature a unique, flexible, over-locking design, locking securely on the outside of the tube lip and can be removed with one hand. A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples. This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus. They are colour coded for easy specimen identification and to help guard against cross-contamination. The top of each cap can also be numbered or labelled with indelible ink. Self-locking cap holds firmly in any position and keeps contents secure while in transit. Liquid-tight seal withstands centrifugation, agitation, refrigeration and freezing.
  • Colour coded self-locking test tube caps prevent spillage or loss of samples
  • Fit all varieties of blood collecting and disposable culture tubes
Balení: Caps are packed in bags of 1000.