Tubing, sanitary, Tygon® 3350
Supplier: Saint Gobain Life Sciences
070497-09 070497-11 070497-14 070497-16 070498-20 070497-13F 070497-20F 070498-18
224-2538EA 103 EUR
224-2538 224-2542 224-2546 224-2549 224-2523 224-0140 224-0142 224-2521
Tubing, sanitary, Tygon® 3350
Tubing Transfer Tubing
Silicone, platinum cured, transparent.
  • Biocompatible, ideal for sensitive applications, can be used for foodstuffs
  • Resistant to ozone and UV, autoclavable
Tygon® tubing made from platinum-cured silicone with ultra-smooth surface. High elasticity with good restoring force for constant liquid transfer, even with intermittent pumping action.
Temperature resistance: −80 to +204 °C
Hardness (Shore durometer A): 50
Density: 1,14
Certifikace: FDA approved, compliant with USP class VI.