VWR®, 96-Position Block Systems
Supplier: VWR
VWR®
548-0343EA 509 EUR
548-0343 548-0345 548-0346 548-0349
VWR®, 96-Position Block Systems
Microplates Microplates with Glass Inserts
96-position systems (Ritter) with glass inserts are used in HTS (high throughput screening) chemistry when the pure PP block is not inert enough. The inserts can be sealed with a PE cap seal, or over the whole block with a Sealmat block cover made of silicone, laminated with a PTFE film.