Ubbelohde viscometers for dilution
Supplier: SI Analytics
285401403 285401411 285401428 285401444 285401452
SCOI285401403EA 426 EUR
SCOI285401403 616-0031 SCOI285401428 SCOI285401444 SCOI285401452
Ubbelohde viscometers for dilution
Ubbelohde viscometers with suspended ball level used to determine the limit viscosity of polymers.
The limit number is determined in combination with the viscosity measuring instruments and piston burette.