Roll dispensers for centrefeed, 7087
Supplier: Kimberly-Clark
115-2182EA 133 EUR
115-2182
Roll dispensers for centrefeed, 7087
Wipe Dispensers
Dispenser made from durable plastic. For use with WYPALL* L10, L20, L30 wipes.
  • Dispenses wipes individually
  • Easy to refill
  • Convenient and low cost