Scrubbers for Kjeldahl mineralisation units
Supplier: Gerhardt
Turbosog
705-0488EA 0 EUR
705-0488
Scrubbers for Kjeldahl mineralisation units
Distillation Systems Scrubber Systems
The Turbosog scubber unit condenses and neutralises aggressive acid vapours from the Kjeldahl digestion, including perchloric acid and other toxic gases.
  • Pre-separator has two bottles for separation and washing
  • Water consumption of 50 L/h
  • Unit is easy to move