Universal upright freezers, GP models
Supplier: Liebherr
LIEBGP1486EA 0 EUR
LIEBGP1486 LIEBGP1476 471-1068 LIEBGP1213 471-0414 LIEBGP1376X 471-1067 LIEBGP2733X LIEBGP2433X LIEBGP1213X 471-1021
Universal upright freezers, GP models
Freezers Laboratory / General Purpose Freezers
These are general purpose freezers with static cooling - ideal for the storage of non critical products. Spacious, energy-saving, these upright freezers are highly economical with optimised cooling systems and compressors. They use very little electricity and are very user friendly. All models are equipped with a recovery defrost water system. All models include SuperFrost technology which makes freezing simple and saves energy, it quickly reduces the temperature to −32 °C.
  • Electronic temperature control
  • All drawers can be removed to increase storage space
Temperature range: −18 to − 25 °C