Vertical laminar flow product protection workbenches, DLF
Supplier: Telstar
135-1509EA 0 EUR
CLEAP0026360 CLEAP0026660
Vertical laminar flow product protection workbenches, DLF
The versatile DLF series is suitable for working with non hazardous materials that require a low-particle working environment. For use in the pharmaceutical industry, biotechnology, hospitals, university and scientific laboratories as well as cell and tissue culture laboratories. The highest quality materials are used to ensure very quiet running, the noise level is 58 dB (A). Integral alarm function (visual and audible).
  • Cleanroom Class 100 (ISO 5)
  • Impact resistant housing made from epoxy-coated steel
  • The pre-filter and HEPA filter are easy to replace
  • Microprocessor control display with electronic Flow Control shows all important data at a glance
DLF 360 PCR is fitted with a stainless steel worktop, folding acrylic window, UV lamp and operating hours counter as standard. The standard UV lamp reliably protects against cross-contamination by nucleic acid. The folding PC window is resistant to UV.
Certifikace: EN 1822 (HEPA filter type H14); EN 779 (pre-filter type Type G4)
Delivery information: Delivery prices available upon request. Installation and commissioning prices available on request.