Gas cylinder holder
Supplier: Usbeck
159-9110EA 53.8 EUR
159-9110
Gas cylinder holder
Wall Hooks
Powder coated aluminium gas cylinder holder with safety belt and snap buckles. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 70 mm thickness.
  • Suitable for all cylinders size 10 litres and above
  • Secures tightly, can be loosened quickly