Orbital shakers, Vibramax
Supplier: Heidolph
Vibramax
444-1487EA 918 EUR
444-1487 444-1220 444-1231 444-1232 444-1230
Orbital shakers, Vibramax
Shakers Orbital Shakers
Vibrating platform shakers designed for test tubes or other vessels with analogue control dials for vibration frequency. The insulated motor ensures virtually no heat transfer from motor to platform.
  • Continuous or timer operation: Allows for unattended operation up to 120 min, sends an acoustic signal
  • Vibramax 100 has larger orbit for best mixing results even with small quantities
  • Smaller orbit of Vibramax 110 prevents contamination even with full tubes
  • Can be used at 0 to 50 °C (80% relative humidity), in an incubator or cold room
IP Protection Class according to DIN EN 60529: IP 30
Certifikace: IP Protection Class according to DIN EN 60529: IP 30
Informace pro objednávání: Supplied with non-slip rubber mat, required accessories must be ordered separately. Vibramax 100 accessories are category B and Vibramax 110 accessories are category C.