Mesh
Supplier: NEOLAB
4-1403 1-2000 1-1600 1-1000 1-900 1-800 4-3300 4-2000 4-1000 4-900 3-3360 3-2400 3-2000 3-1200 3-1000 1-710 1-600 1-500 1-400 1-300 1-250 1-200 1-150 1-105 1-90 1-80 1-70 1-53 1-40 1-30 1-20 1-10 4-700 4-600 4-500 4-400 4-300 4-250 4-200 4-150 4-125 4-90 4-80 4-70 4-60 4-50 4-40 4-30 4-20 4-15 3-710 3-500 3-350 3-295 3-210 3-150 2-70 2-85 2-150 2-210 2-300 2-420 2-500 2-590 2-850 2-1000 4-5 4-1180 4-1 1-5 4-56 4-1550 4-1800 4-25 4-350 4-10 4-140
510-9483EA 87.4 EUR
510-9483 510-9470 510-9471 510-9472 510-9473 510-9474 510-9481 510-9482 510-9484 510-9485 510-9490 510-9491 510-9492 510-9494 510-9495 510-9501 510-9502 510-9503 510-9504 510-9505 510-9506 510-9507 510-9508 510-9509 510-9510 510-9511 510-9512 510-9513 510-9514 510-9515 510-9516 510-9517 510-9521 510-9522 510-9523 510-9524 510-9525 510-9526 510-9527 510-9528 510-9529 510-9531 510-9532 510-9533 510-9534 510-9535 510-9536 510-9537 510-9538 510-9539 510-9551 510-9552 510-9553 510-9554 510-9556 510-9557 510-9561 510-9562 510-9568 510-9569 510-9574 510-9575 510-9576 510-9577 510-9578 510-9579 510-2076 510-2068 510-2066 STOC1-5 510-2077 510-2070 510-2071 510-2073 510-2074 510-2067 510-2069
Mesh
Filters Membrane Filters
Polyamide mesh (Nylon): Resistance to acids is limited and dependent on the concentration and temperature. Excellent resistance to alkalis, including caustic soda and caustic potash. Chemical performance against organic solvents is very good. Temperature resistant from 0 to 130 °C when dry, 95 to 100 °C in water.
Polyester mesh: Resistant to most acids. Not resistant to alkalis. Temperature resistant from –75 to +150 °C (for short periods), up to 125 °C for longer requirements.
PP mesh: Humidity resolution at 95% rel. air moisture and 25 °C is less than 0,01%. Resistant to alkalis, even in high concentrations. Use with organic solvents is limited. Temperature resistant from 0 to +150 °C (for short periods), up to 125 °C for longer requirements.
ETFE mesh: Humidity absorption at 20 °C and 65% relative humidity is 0%. Good resistance to acids, alkalis, solvents and hydrolysis. Temperature resistant from –190 to +150 °C when dry.