VWR® VHP-C, Hotplates
Supplier: VWR Collection
453-0050EA 559 EUR
453-0050 453-0051 453-0053 453-0054 453-0056 453-0057
VWR® VHP-C, Hotplates
Heaters Hotplates
Compact hotplates with a chemically resistant ceramic top. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature a digital error code display and 'hot' top indicator to warn users that the surface is hot.
  • VHP-C7 can be used with the VT-5 contact thermometer (442-0573), enabling precise temperature control
  • Fixed safety circuit shuts off heater if temperature rises above 550 °C
  • Microprocessor control sends feedback to the hotplate to maintain set temperature
  • Digital display ensures accurate and repeatable temperature settings
IP protection class according to DIN EN 60529: IP 21