Stirring paddles, polypropylene
Supplier: KARTELL
428 430 431 432
442-4100EA 45.3 EUR
442-4100 441-9511 442-4120 442-2121
Stirring paddles, polypropylene
Stirrers Stirring Tools
Blades are made of PP, and are fixed to a PP covered steel rod.
  • Autoclavable
  • Good chemical resistance