Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
0739000 0741000 0741300
441-2904EA 786 EUR
441-2904 441-2901 441-2903
Overhead stirrer tools, 4-blade stirrers
Stirrers Stirring Tools
Made from stainless steel. Stirrers draw the material to be mixed from the top of the vessel to the bottom. Local shearing forces.
  • For medium to high speed range
  • Ideal for stirring liquids with low to medium viscosity
  • Creates axial flow