Overhead stirrer tools, dissolver stirrers
Supplier: IKA
0513500 2387900 2746700
441-2951EA 324 EUR
441-2951 441-2953 441-2954
Overhead stirrer tools, dissolver stirrers
Stirrers Stirring Tools
Made from stainless steel. Stirrers draw the material to be mixed from the top and the bottom. High turbulence and high shearing forces.
  • Ideal for stirring liquids at medium to high speeds
  • Creates radial flow
  • Choice of three different stirrer formats