TLC/HPTLC plates developing system, smartAlert
Supplier: CAMAG
smartAlert
552-0078EA 389 EUR
552-0078
TLC/HPTLC plates developing system, smartAlert
TLC Plate Monitors
Ideal solution for monitoring the solvent front in development of TLC/HPTLC plates without relying on a stopwatch. The desired developing distance is set, and as soon as the solvent front has reached this position, visual and acoustic signals alert the operator. This allows more constructive and efficient time/tasks management as well as guaranteeing accurate results. Designed to work with TLC/HPTLC glass chambers and plates up to 200×200 mm, battery powered (up to 1000 developments).
  • Improved reproducibility of results
  • Easy to operate
  • Cost and time efficient, no waste of over-developed plates